Assay Details
| Gene Symbol: Ddx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145885.1 | NP_001139357.1 | 9 - 10 | 633 | 145 | |
| NM_010029.2 | NP_034159.1 | 8 - 9 | 555 | 145 | ||
| GenBank mRNA | AK014844.1 | - | 8 - 9 | 546 | 145 | |
| AK133366.1 | - | 9 - 10 | 626 | 145 | ||
| D14859.1 | - | 5 - 6 | 328 | 145 | ||
More Information
Set Membership: |
|

Ddx4
Genomic Map