Assay Details
| Gene Symbol: Doc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010069.1 | NP_034199.1 | 3 - 4 | 1006 | 89 | |
| GenBank mRNA | BC055768.1 | - | 3 - 4 | 1006 | 6413208 | 89 |
| D50000.1 | - | 2 - 3 | 429 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Doc2a
Genomic Map