Assay Details
| Gene Symbol: Adgre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010130.4 | NP_034260.1 | 4 - 5 | 434 | 70 | |
| GenBank mRNA | AK133202.1 | - | 4 - 5 | 410 | 70 | |
| AK151303.1 | - | 4 - 5 | 406 | 70 | ||
| AK151682.1 | - | 4 - 5 | 426 | 70 | ||
| AK155862.1 | - | 4 - 5 | 385 | 70 | ||
| BC075688.1 | - | 4 - 5 | 406 | 30609706 | 70 | |
| U66888.1 | - | 4 - 5 | 421 | 70 | ||
| U66889.1 | - | 4 - 5 | 421 | 70 | ||
| U66890.1 | - | 4 - 5 | 421 | 70 | ||
| X93328.1 | - | 4 - 5 | 430 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre1
Genomic Map