Assay Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271962.1 | NP_001258891.1 | 9 - 10 | 944 | 58 | |
| NM_001271963.1 | NP_001258892.1 | 9 - 10 | 1120 | 58 | ||
| NM_010165.3 | NP_034295.1 | 9 - 10 | 1121 | 58 | ||
| GenBank mRNA | AK137718.1 | - | 9 - 10 | 1116 | 58 | |
| AK141765.1 | - | 9 - 10 | 1120 | 58 | ||
| AY212511.1 | - | 6 - 7 | 635 | 58 | ||
| BC003755.1 | - | 9 - 10 | 1121 | 3497245 | 58 | |
| U61111.1 | - | 8 - 9 | 862 | 58 | ||
| U71208.1 | - | 9 - 10 | 946 | 58 | ||
| U81603.1 | - | 8 - 9 | 861 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya2
Genomic Map