Assay Details
| Gene Symbol: Fh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010209.2 | NP_034339.2 | 1 - 2 | 177 | 95 | |
| GenBank mRNA | AK002379.1 | - | 1 - 2 | 129 | 95 | |
| AK147000.1 | - | 1 - 2 | 171 | 95 | ||
| BC006048.1 | - | 1 - 2 | 120 | 3590691 | 95 | |
| BU526396.1 | - | 1 - 2 | 180 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fh1
Genomic Map