Assay Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010236.2 | NP_034366.2 | 11 - 12 | 1112 | 69 | |
| GenBank mRNA | AK041952.1 | - | 9 - 10 | 1254 | 69 | |
| AK157040.1 | - | 11 - 12 | 1073 | 69 | ||
| BC005484.1 | - | 11 - 12 | 1055 | 3485401 | 69 | |
| BM944126.1 | - | 2 - 3 | 111 | 69 | ||
| CF749232.1 | - | 8 - 9 | 711 | 69 | ||
| U32197.1 | - | 11 - 12 | 1069 | 69 | ||
| U33557.1 | - | 11 - 12 | 1089 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fpgs
Genomic Map