Assay Details
| Gene Symbol: Gata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010258.3 | NP_034388.2 | 5 - 6 | 1772 | 100 | |
| GenBank mRNA | AF179425.1 | - | 4 - 5 | 1079 | 100 | |
| AK142381.1 | - | 5 - 6 | 1632 | 100 | ||
| BG079878.2 | - | 1 - 2 | 104 | 100 | ||
| S82462.1 | - | 4 - 5 | 1534 | 100 | ||
| U51335.1 | - | 3 - 4 | 352 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gata6
Genomic Map