Assay Details
| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010261.2 | NP_034391.1 | 4 - 5 | 616 | 60 | |
| GenBank mRNA | AF120162.1 | - | 4 - 5 | 560 | 60 | |
| AF252856.1 | - | 4 - 5 | 550 | 60 | ||
| AK008559.1 | - | 4 - 5 | 528 | 60 | ||
| AK009726.1 | - | 4 - 5 | 476 | 60 | ||
| AK046109.1 | - | 3 - 4 | 512 | 60 | ||
| AK150681.1 | - | 4 - 5 | 497 | 60 | ||
| AK170289.1 | - | 4 - 5 | 576 | 60 | ||
| BC008242.1 | - | 4 - 5 | 549 | 2655184 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007634 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007636 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007550 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043723 | AK009726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043723 | AK046109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043723 | AK150681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043723 | AK170289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043723 | BC008242.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955227 | AK009726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955227 | AK046109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955227 | AK150681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955227 | AK170289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955227 | BC008242.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80368 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155406 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043723 | AK009726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043723 | AK046109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043723 | AK150681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043723 | AK170289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043723 | BC008242.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955227 | AK009726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955227 | AK046109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955227 | AK150681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955227 | AK170289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955227 | BC008242.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25007645 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155406 | - | - |
| Whole Mouse Genome G4122A | A_51_P374383 | AF252856.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111099 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06344668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06344670 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06098563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06098579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06098578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007551 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007634 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007636 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25007549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05955756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05955752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06076709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06077040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06128910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06344670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05985980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06098572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06076706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25007606 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560912 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427773_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560911 | - | - |
| Mouse Genome 430 2.0 Array | 1427773_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427773_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14470_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14470_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022547.mm.1 | TC0700002615.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022547.mm.1 | NM_010261.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022547.mm.1 | NM_010261.2 | Refseq |
| Mouse Expression Set 430 | 1427773_a_at | - | - |
| Mouse Expression Set 430 | 1427773_PM_a_at | - | - |
| Murine Genome U74v2 Set | 94269_at | - | - |
| Murine Genome U74v2 Set | 94269_PM_at | - | - |
| Murine Genome U74v2 Set | 167883_at | - | - |
| Murine Genome U74v2 Set | 167883_PM_at | - | - |
| Murine Genome U74v2 Set | 168504_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168504_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427773_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427773_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427773_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427773_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2030435 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2100632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030435 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244857 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2710113 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2710113 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244857 | - | - |
| mouse-6 v1.1 (Illumina) | scl00015.1_21-S | - | - |
| mouse-6 v1.1 (Illumina) | scl014470.4_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100632 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014470.4_28-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00015.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100632 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2710112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244857 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2710112 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244857 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014470.4_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00015.1_21-S | - | - |

Rabac1
Genomic Map