Assay Details
| Gene Symbol: Igf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010513.2 | NP_034643.2 | 16 - 17 | 3795 | 70 | |
| GenBank mRNA | AF056187.1 | - | 16 - 17 | 3193 | 70 | |
| AK137220.1 | - | 11 - 12 | 1924 | 70 | ||
| AK155884.1 | - | 15 - 16 | 2691 | 70 | ||
| BC138868.1 | - | 16 - 17 | 3225 | 8861890 | 70 | |
| BC138869.1 | - | 16 - 17 | 3225 | 8861891 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf1r
Genomic Map