Assay Details
| Gene Symbol: Igf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010513.2 | NP_034643.2 | 7 - 8 | 2196 | 69 | |
| GenBank mRNA | AF056187.1 | - | 7 - 8 | 1594 | 69 | |
| AK137220.1 | - | 2 - 3 | 325 | 69 | ||
| AK155079.1 | - | 6 - 7 | 1074 | 69 | ||
| AK155884.1 | - | 6 - 7 | 1092 | 69 | ||
| BC138868.1 | - | 7 - 8 | 1626 | 8861890 | 69 | |
| BC138869.1 | - | 7 - 8 | 1626 | 8861891 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf1r
Genomic Map