Assay Details
| Gene Symbol: Inppl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122739.1 | NP_001116211.1 | 9 - 10 | 1245 | 65 | |
| NM_010567.2 | NP_034697.2 | 9 - 10 | 1479 | 65 | ||
| GenBank mRNA | AF162781.1 | - | 9 - 10 | 1329 | 65 | |
| AK133359.1 | - | 9 - 10 | 1206 | 65 | ||
| AK149873.1 | - | 9 - 10 | 1263 | 65 | ||
| AK216836.1 | - | 3 - 4 | 252 | 65 | ||
| BC063080.1 | - | 9 - 10 | 1175 | 6416946 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inppl1
Genomic Map