Assay Details
| Gene Symbol: Nfatc2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010900.3 | NP_035030.2 | 6 - 7 | 991 | 71 | |
| GenBank mRNA | AK005947.1 | - | 6 - 7 | 982 | 71 | |
| AK030107.1 | - | 6 - 7 | 993 | 71 | ||
| BC113761.1 | - | 6 - 7 | 992 | 40054445 | 71 | |
| U76759.1 | - | 6 - 7 | 981 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc2ip
Genomic Map