Assay Details
| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098404.1 | NP_001091874.1 | 6 - 7 | 1141 | 79 | |
| NM_010936.3 | NP_035066.1 | 6 - 7 | 1264 | 79 | ||
| GenBank mRNA | AF031814.1 | - | 6 - 7 | 1079 | 79 | |
| AK018630.1 | - | 6 - 7 | 1264 | 79 | ||
| BC120796.1 | - | 6 - 7 | 1011 | 40129719 | 79 | |
| BC137780.1 | - | 6 - 7 | 1011 | 8860799 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr1i2
Genomic Map