Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 11 - 12 | 1514 | 68 | |
| GenBank mRNA | AK030574.1 | - | 11 - 12 | 1443 | 68 | |
| AK045008.1 | - | 11 - 12 | 1443 | 68 | ||
| AK149127.1 | - | 11 - 12 | 1406 | 68 | ||
| AK160798.1 | - | 11 - 12 | 1443 | 68 | ||
| AK161212.1 | - | 10 - 11 | 1207 | 68 | ||
| AK165468.1 | - | 11 - 12 | 1443 | 68 | ||
| AK169726.1 | - | 11 - 12 | 1447 | 68 | ||
| BC009654.1 | - | 11 - 12 | 1437 | 3484815 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

P4ha1
Genomic Map