Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 13 - 14 | 1647 | 67 | |
| GenBank mRNA | AK030574.1 | - | 13 - 14 | 1576 | 67 | |
| AK045008.1 | - | 13 - 14 | 1576 | 67 | ||
| AK149127.1 | - | 13 - 14 | 1539 | 67 | ||
| AK160798.1 | - | 13 - 14 | 1576 | 67 | ||
| AK161212.1 | - | 12 - 13 | 1340 | 67 | ||
| AK165468.1 | - | 13 - 14 | 1576 | 67 | ||
| AK169726.1 | - | 13 - 14 | 1580 | 67 | ||
| BC009654.1 | - | 13 - 14 | 1570 | 3484815 | 67 | |
| U16162.1 | - | 12 - 13 | 1417 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha1
Genomic Map