Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 4 - 5 | 531 | 75 | |
| GenBank mRNA | AK030574.1 | - | 4 - 5 | 463 | 75 | |
| AK045008.1 | - | 4 - 5 | 460 | 75 | ||
| AK149127.1 | - | 4 - 5 | 423 | 75 | ||
| AK160798.1 | - | 4 - 5 | 460 | 75 | ||
| AK161212.1 | - | 4 - 5 | 464 | 75 | ||
| AK165468.1 | - | 4 - 5 | 460 | 75 | ||
| AK169726.1 | - | 4 - 5 | 464 | 75 | ||
| BC009654.1 | - | 4 - 5 | 454 | 3484815 | 75 | |
| BY721363.1 | - | 4 - 5 | 512 | 75 | ||
| U16162.1 | - | 3 - 4 | 301 | 75 | ||
More Information
Set Membership: |
|

P4ha1
Genomic Map