Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 6 - 7 | 910 | 119 | |
| GenBank mRNA | AK045008.1 | - | 6 - 7 | 839 | 119 | |
| AK149127.1 | - | 6 - 7 | 802 | 119 | ||
| AK160798.1 | - | 6 - 7 | 839 | 119 | ||
| AK165468.1 | - | 6 - 7 | 839 | 119 | ||
| AK169726.1 | - | 6 - 7 | 843 | 119 | ||
| BC009654.1 | - | 6 - 7 | 833 | 3484815 | 119 | |
| U16162.1 | - | 5 - 6 | 680 | 119 | ||
More Information
Set Membership: |
|

P4ha1
Genomic Map