Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011030.2 | NP_035160.1 | 8 - 9 | 1281 | 78 | |
| GenBank mRNA | AK030574.1 | - | 8 - 9 | 1211 | 78 | |
| AK149127.1 | - | 8 - 9 | 1173 | 78 | ||
| AK160798.1 | - | 8 - 9 | 1210 | 78 | ||
| AK165468.1 | - | 8 - 9 | 1210 | 78 | ||
| AK169726.1 | - | 8 - 9 | 1214 | 78 | ||
| BC009654.1 | - | 8 - 9 | 1204 | 3484815 | 78 | |
More Information
Set Membership: |
|

P4ha1
Genomic Map