Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024955.2 | NP_001020126.1 | 4 - 5 | 836 | 160 | |
| NM_001077495.2 | NP_001070963.1 | 10 - 11 | 1925 | 160 | ||
| GenBank mRNA | AK046259.1 | - | 4 - 5 | 838 | 161 | |
| BC026146.1 | - | 10 - 11 | 1906 | 3979333 | 160 | |
| BC051106.1 | - | 4 - 5 | 536 | 5031261 | 160 | |
| CB519962.1 | - | 4 - 5 | 468 | 160 | ||
| M60651.1 | - | 10 - 11 | 1860 | 160 | ||
| U50413.1 | - | 10 - 11 | 1878 | 160 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map