Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024955.2 | NP_001020126.1 | 5 - 6 | 955 | 163 | |
| NM_001077495.2 | NP_001070963.1 | 11 - 12 | 2044 | 163 | ||
| GenBank mRNA | AK046259.1 | - | 5 - 6 | 958 | 164 | |
| BC026146.1 | - | 11 - 12 | 2025 | 3979333 | 163 | |
| BC051106.1 | - | 5 - 6 | 655 | 5031261 | 163 | |
| CB519962.1 | - | 5 - 6 | 587 | 163 | ||
| M60651.1 | - | 11 - 12 | 1979 | 163 | ||
| U50413.1 | - | 11 - 12 | 1997 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map