Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024955.2 | NP_001020126.1 | 7 - 8 | 1277 | 73 | |
| NM_001077495.2 | NP_001070963.1 | 13 - 14 | 2366 | 73 | ||
| GenBank mRNA | AK046259.1 | - | 7 - 8 | 1280 | 73 | |
| BC026146.1 | - | 13 - 14 | 2347 | 3979333 | 73 | |
| BC051106.1 | - | 7 - 8 | 977 | 5031261 | 73 | |
| M60651.1 | - | 13 - 14 | 2301 | 73 | ||
| U50413.1 | - | 13 - 14 | 2319 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map