Assay Details
| Gene Symbol: Ppp5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011155.2 | NP_035285.2 | 11 - 12 | 1458 | 70 | |
| GenBank mRNA | AF018262.1 | - | 11 - 12 | 1429 | 70 | |
| BC003744.1 | - | 11 - 12 | 1414 | 3590322 | 70 | |
| CD775986.1 | - | 2 - 3 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp5c
Genomic Map