Assay Details
| Gene Symbol: Ppp5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011155.2 | NP_035285.2 | 8 - 9 | 1147 | 69 | |
| GenBank mRNA | AF018262.1 | - | 8 - 9 | 1118 | 69 | |
| AK189512.1 | - | 1 - 2 | 31 | 69 | ||
| BC003744.1 | - | 8 - 9 | 1103 | 3590322 | 69 | |
More Information
Set Membership: |
|

Ppp5c
Genomic Map