Assay Details
| Gene Symbol: Psmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011188.3 | NP_035318.1 | 2 - 3 | 466 | 72 | |
| GenBank mRNA | AK005083.1 | - | 1 - 2 | 89 | 72 | |
| AK076874.1 | - | 2 - 3 | 468 | 72 | ||
| AK146919.1 | - | 1 - 2 | 107 | 72 | ||
| AK153413.1 | - | 1 - 2 | 110 | 72 | ||
| AK168548.1 | - | 1 - 2 | 107 | 72 | ||
| BC005462.1 | - | 1 - 2 | 116 | 3590739 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc2
Genomic Map