Assay Details
| Gene Symbol: Psmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011188.3 | NP_035318.1 | 5 - 6 | 689 | 81 | |
| GenBank mRNA | AK005083.1 | - | 4 - 5 | 312 | 81 | |
| AK076874.1 | - | 5 - 6 | 691 | 81 | ||
| AK146919.1 | - | 4 - 5 | 330 | 81 | ||
| AK168548.1 | - | 4 - 5 | 330 | 81 | ||
| AK216249.1 | - | 1 - 2 | 62 | 81 | ||
| BC005462.1 | - | 4 - 5 | 339 | 3590739 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc2
Genomic Map