Assay Details
| Gene Symbol: Psmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011188.3 | NP_035318.1 | 7 - 8 | 897 | 65 | |
| GenBank mRNA | AK005083.1 | - | 6 - 7 | 520 | 65 | |
| AK076874.1 | - | 7 - 8 | 899 | 65 | ||
| AK146919.1 | - | 6 - 7 | 538 | 65 | ||
| AK153413.1 | - | 6 - 7 | 541 | 65 | ||
| AK168548.1 | - | 6 - 7 | 538 | 65 | ||
| BC005462.1 | - | 6 - 7 | 547 | 3590739 | 65 | |
| U61283.1 | - | 6 - 7 | 583 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc2
Genomic Map