Assay Details
| Gene Symbol: Pstpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088150.1 | - | 11 - 12 | 2167 | 74 | |
| U87814.1 | - | 12 - 13 | 1205 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pstpip1| Gene Symbol: Pstpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088150.1 | - | 11 - 12 | 2167 | 74 | |
| U87814.1 | - | 12 - 13 | 1205 | 74 | ||
Set Membership: |
|