Assay Details
| Gene Symbol: Ptprn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011215.2 | NP_035345.2 | 18 - 19 | 2714 | 75 | |
| GenBank mRNA | AK138655.1 | - | 18 - 19 | 2706 | 75 | |
| BC133678.1 | - | 18 - 19 | 2598 | 40109882 | 75 | |
| U57345.1 | - | 18 - 19 | 2601 | 75 | ||
| U82439.1 | - | 13 - 14 | 1722 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprn2
Genomic Map