Assay Details
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011226.1 | NP_035356.1 | 2 - 3 | 301 | 89 | |
GenBank mRNA | AB232613.1 | - | 1 - 2 | 207 | 89 | |
AK017590.1 | - | 2 - 3 | 335 | 89 | ||
BC032936.1 | - | 2 - 3 | 301 | 1379043 | 89 | |
X80473.1 | - | 1 - 2 | 207 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
