| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900011761.mm.1 |
NM_011265.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011761.mm.1 |
NM_011265.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011761.mm.1 |
TC1900001316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011760.mm.1 |
NM_011265.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011760.mm.1 |
NM_011265.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011760.mm.1 |
TC1900001316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011751.mm.1 |
NM_001166414.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011751.mm.1 |
NM_001166414.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011751.mm.1 |
NM_011265.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011751.mm.1 |
NM_011265.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011751.mm.1 |
TC1900001316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900011758.mm.1 |
NM_011265.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011758.mm.1 |
NM_011265.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900011758.mm.1 |
TC1900001316.mm.1 |
MTA TC |