Assay Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166414.1 | NP_001159886.1 | 7 - 8 | 892 | 108 | |
| NM_011265.3 | NP_035395.2 | 6 - 7 | 933 | 108 | ||
| GenBank mRNA | AK030008.1 | - | 8 - 9 | 965 | 108 | |
| AK041120.1 | - | 6 - 7 | 918 | 108 | ||
| AK132267.1 | - | 2 - 3 | 258 | 108 | ||
| AK140928.1 | - | 5 - 6 | 742 | 108 | ||
| BC017598.1 | - | 6 - 7 | 907 | 4483833 | 108 | |
| X76090.1 | - | 3 - 4 | 324 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx3
Genomic Map