Assay Details
| Gene Symbol: Kdm5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011419.3 | NP_035549.1 | 14 - 15 | 2236 | 76 | |
| GenBank mRNA | AB221607.1 | - | 14 - 15 | 2062 | 76 | |
| AF127244.1 | - | 14 - 15 | 2074 | 76 | ||
| BC059077.1 | - | 6 - 7 | 1051 | 6826863 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0Y00000005.rn
Set Membership: |
|

Kdm5d
Genomic Map