Assay Details
| Gene Symbol: Tgfbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011578.3 | NP_035708.2 | 2 - 3 | 498 | 88 | |
| GenBank mRNA | AF039601.1 | - | 2 - 3 | 298 | 88 | |
| AK165763.1 | - | 2 - 3 | 415 | 88 | ||
| AK166082.1 | - | 2 - 3 | 499 | 88 | ||
| BC070428.1 | - | 2 - 3 | 423 | 6410554 | 88 | |
More Information
Set Membership: |
|

Tgfbr3
Genomic Map