Assay Details
| Gene Symbol: Dido1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291432.1 | NP_001278361.1 | 4 - 5 | 1425 | 79 | |
| NM_001291433.1 | NP_001278362.1 | 4 - 5 | 1477 | 79 | ||
| NM_011805.3 | NP_035935.2 | 4 - 5 | 1477 | 79 | ||
| NM_175551.4 | NP_780760.2 | 4 - 5 | 1477 | 79 | ||
| NM_177852.4 | NP_808520.2 | 4 - 5 | 1425 | 79 | ||
| NR_111965.1 | 3 - 4 | 1335 | 79 | |||
| GenBank mRNA | AJ238332.1 | - | 4 - 5 | 1379 | 79 | |
| AK042474.1 | - | 4 - 5 | 1446 | 79 | ||
| AK129117.1 | - | 4 - 5 | 1425 | 79 | ||
| AY425951.1 | - | 4 - 5 | 1379 | 79 | ||
| AY425952.1 | - | 4 - 5 | 1379 | 79 | ||
| BC029110.1 | - | 5 - 6 | 1479 | 79 | ||
| BC060249.1 | - | 4 - 5 | 1425 | 79 | ||
| BC096662.1 | - | 4 - 5 | 1417 | 79 | ||
| BC138712.1 | - | 3 - 4 | 1254 | 8861734 | 79 | |
| BC138713.1 | - | 3 - 4 | 1254 | 8861735 | 79 | |
| BC145401.1 | - | 3 - 4 | 1255 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dido1
Genomic Map