Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 14 - 15 | 2339 | 73 | |
| NM_011815.5 | NP_035945.1 | 15 - 16 | 2477 | 73 | ||
| GenBank mRNA | AF001863.1 | - | 14 - 15 | 2315 | 73 | |
| AF061744.1 | - | 15 - 16 | 2453 | 73 | ||
| AK038225.1 | - | 14 - 15 | 2367 | 73 | ||
| AK137907.1 | - | 14 - 15 | 2295 | 73 | ||
| AK172164.1 | - | 8 - 9 | 709 | 73 | ||
| BC141352.1 | - | 15 - 16 | 2418 | 9055964 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fyb
Genomic Map