Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 15 - 16 | 1475 | 98 | |
| GenBank mRNA | AJ006130.1 | - | 15 - 16 | 1358 | 98 | |
| AK082356.1 | - | 15 - 16 | 1407 | 98 | ||
| AK087976.1 | - | 15 - 16 | 1421 | 98 | ||
| BC006706.1 | - | 13 - 14 | 1833 | 98 | ||
| BC014739.1 | - | 15 - 16 | 1406 | 4920184 | 98 | |
| BC026569.1 | - | 15 - 16 | 1406 | 4982612 | 98 | |
| BC029148.1 | - | 13 - 14 | 2369 | 98 | ||
| BC029671.1 | - | 15 - 16 | 1418 | 5364953 | 98 | |
| BC029694.1 | - | 15 - 16 | 1415 | 5369477 | 98 | |
| BC033356.1 | - | 15 - 16 | 1402 | 5366230 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map