Assay Details
| Gene Symbol: Pmm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282040.1 | NP_001268969.1 | 3 - 4 | 325 | 62 | |
| NM_013872.4 | NP_038900.1 | 4 - 5 | 505 | 62 | ||
| GenBank mRNA | AF007267.1 | - | 4 - 5 | 376 | 62 | |
| AK004631.1 | - | 4 - 5 | 438 | 62 | ||
| AK013805.1 | - | 4 - 5 | 409 | 62 | ||
| AK166999.1 | - | 4 - 5 | 440 | 62 | ||
| BC006809.1 | - | 3 - 4 | 264 | 3153981 | 62 | |
| CX217041.1 | - | 4 - 5 | 505 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmm1
Genomic Map