Assay Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020601.2 | NP_065626.1 | 14 - 15 | 1645 | 80 | |
| GenBank mRNA | AK030547.1 | - | 14 - 15 | 1860 | 80 | |
| AK031937.1 | - | 10 - 11 | 1106 | 80 | ||
| AK036996.1 | - | 11 - 12 | 3476 | 80 | ||
| AK038674.1 | - | 14 - 15 | 1646 | 80 | ||
| AK215750.1 | - | 3 - 4 | 355 | 80 | ||
| BC043105.1 | - | 14 - 15 | 1768 | 6406579 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1x
Genomic Map