Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 17 - 18 | 1517 | 81 | |
| NM_198438.1 | NP_940840.1 | 16 - 17 | 1436 | 81 | ||
| GenBank mRNA | AF170906.1 | - | 16 - 17 | 1166 | 81 | |
| AF500117.1 | - | 16 - 17 | 1166 | 81 | ||
| AK011509.1 | - | 17 - 18 | 1132 | 81 | ||
| AK011853.1 | - | 17 - 18 | 1487 | 81 | ||
| AK017711.1 | - | 17 - 18 | 81 | |||
| AK041388.1 | - | 13 - 14 | 1033 | 81 | ||
| AK133812.1 | - | 14 - 15 | 976 | 81 | ||
| AK153733.1 | - | 13 - 14 | 877 | 81 | ||
| AY167987.1 | - | 16 - 17 | 1049 | 81 | ||
| BC003430.1 | - | 16 - 17 | 1190 | 2646149 | 81 | |
| BC028324.1 | - | 11 - 12 | 658 | 81 | ||
| BC035475.1 | - | 16 - 17 | 1379 | 81 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map