Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162533.1 | NP_001156005.1 | 13 - 14 | 1127 | 113 | |
| NM_025856.3 | NP_080132.1 | 9 - 10 | 791 | 113 | ||
| GenBank mRNA | AK011430.1 | - | 8 - 9 | 1112 | 113 | |
| AK155132.1 | - | 13 - 14 | 1048 | 113 | ||
| BC019568.1 | - | 13 - 14 | 1044 | 113 | ||
| BC025637.1 | - | 7 - 8 | 788 | 113 | ||
| BC039777.1 | - | 9 - 10 | 981 | 113 | ||
| BC053421.1 | - | 9 - 10 | 752 | 6706860 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3d21
Genomic Map