Assay Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271397.1 | NP_001258326.1 | 4 - 5 | 909 | 83 | |
| NR_073167.1 | 4 - 5 | 1090 | 83 | |||
| NR_073168.1 | 4 - 5 | 905 | 83 | |||
| GenBank mRNA | AK014994.1 | - | 4 - 5 | 910 | 83 | |
| AK029961.1 | - | 4 - 5 | 622 | 83 | ||
| AK147172.1 | - | 4 - 5 | 645 | 83 | ||
| BC049166.1 | - | 4 - 5 | 500 | 83 | ||
| BC058699.1 | - | 4 - 5 | 1065 | 83 | ||
| BC125575.1 | - | 4 - 5 | 635 | 40130090 | 83 | |
| BC125579.1 | - | 4 - 5 | 635 | 40130094 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nol8
Genomic Map