Assay Details
| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028410.1 | NP_082686.1 | 1 - 2 | 118 | 75 | |
| GenBank mRNA | AK013663.1 | - | 1 - 2 | 209 | 75 | |
| BC049103.1 | - | 1 - 2 | 118 | 6400126 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkrir
Genomic Map