Assay Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029278.2 | NP_083554.2 | 6 - 7 | 964 | 77 | |
| GenBank mRNA | AK011648.1 | - | 6 - 7 | 940 | 77 | |
| AK078605.1 | - | 6 - 7 | 950 | 77 | ||
| AK148605.1 | - | 6 - 7 | 923 | 77 | ||
| AK161728.1 | - | 6 - 7 | 902 | 77 | ||
| BC024998.1 | - | 6 - 7 | 885 | 5012070 | 77 | |
| BC043043.1 | - | 6 - 7 | 907 | 5701430 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nop14
Genomic Map