Assay Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029278.2 | NP_083554.2 | 7 - 8 | 1098 | 158 | |
| GenBank mRNA | AK078605.1 | - | 7 - 8 | 1084 | 158 | |
| AK148605.1 | - | 7 - 8 | 1057 | 158 | ||
| AK161728.1 | - | 7 - 8 | 1036 | 158 | ||
| BC024998.1 | - | 7 - 8 | 1019 | 5012070 | 158 | |
| BC043043.1 | - | 7 - 8 | 1041 | 5701430 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nop14
Genomic Map