Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 19 - 20 | 1658 | 88 | |
| GenBank mRNA | AK089462.1 | - | 19 - 20 | 1682 | 88 | |
| AK133294.1 | - | 19 - 20 | 1800 | 88 | ||
| AK143611.1 | - | 19 - 20 | 1683 | 88 | ||
| AK158367.1 | - | 19 - 20 | 1683 | 88 | ||
| BC052447.1 | - | 19 - 20 | 1658 | 6408647 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930402H24Rik
Genomic Map