Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 23 - 24 | 2089 | 64 | |
| GenBank mRNA | AK133294.1 | - | 23 - 24 | 2231 | 64 | |
| AK143611.1 | - | 23 - 24 | 2114 | 64 | ||
| AK158367.1 | - | 23 - 24 | 2114 | 64 | ||
| BC052447.1 | - | 23 - 24 | 2089 | 6408647 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930402H24Rik
Genomic Map