Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 2 - 3 | 225 | 71 | |
| GenBank mRNA | AK089462.1 | - | 2 - 3 | 249 | 71 | |
| AK133294.1 | - | 2 - 3 | 367 | 71 | ||
| AK143611.1 | - | 2 - 3 | 250 | 71 | ||
| AK158367.1 | - | 2 - 3 | 247 | 71 | ||
| BC052447.1 | - | 2 - 3 | 225 | 6408647 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133294.1 transcripts.
Set Membership: |
|

4930402H24Rik
Genomic Map