Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 9 - 10 | 905 | 82 | |
| GenBank mRNA | AK015050.1 | - | 9 - 10 | 907 | 82 | |
| AK089462.1 | - | 9 - 10 | 929 | 82 | ||
| AK133294.1 | - | 9 - 10 | 1047 | 82 | ||
| AK143611.1 | - | 9 - 10 | 930 | 82 | ||
| AK158367.1 | - | 9 - 10 | 927 | 82 | ||
| BC052447.1 | - | 9 - 10 | 905 | 6408647 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930402H24Rik
Genomic Map