Assay Details
| Gene Symbol: Spint4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030058.2 | NP_084334.1 | 2 - 3 | 357 | 122 | |
| GenBank mRNA | AK020315.1 | - | 2 - 3 | 330 | 122 | |
| AY226988.1 | - | 2 - 3 | 357 | 122 | ||
| BC119299.1 | - | 2 - 3 | 327 | 8734048 | 122 | |
| BC119301.1 | - | 2 - 3 | 327 | 8734050 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spint4
Genomic Map