Assay Details
| Gene Symbol: Loxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033325.2 | NP_201582.2 | 11 - 12 | 2129 | 72 | |
| GenBank mRNA | AF117951.1 | - | 326 | 72 | ||
| AK034957.1 | - | 11 - 12 | 2167 | 72 | ||
| AK045019.1 | - | 11 - 12 | 2129 | 72 | ||
| AK145323.1 | - | 11 - 12 | 2167 | 72 | ||
| AK159386.1 | - | 11 - 12 | 2163 | 72 | ||
| BC086801.1 | - | 2411 | 30605240 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Loxl2
Genomic Map